Learn More
Cayman Chemical Cryogenine, Purity: ≥98%, CAS: 10308-13-1, 5 mg
SDP
Catalog No. 502932596
Encompass_Preferred
An alkaloid with anti-inflammatory activity; inhibits prostaglandin synthetase (IC50 = 424 μM); reduces paw edema and the mean arthritic index in a rat model of adjuvant-induced polyarthritis at 100 mg/kg per day, p.o.
Catalog No. 50-293-2596 Supplier Cayman Chemical Supplier No. 252875 MG
An alkaloid with anti-inflammatory activity; inhibits prostaglandin synthetase (IC50 = 424 μM); reduces paw edema and the mean arthritic index in a rat model of adjuvant-induced polyarthritis at 100 mg/kg per day, p.o.