Learn More
Cayman Chemical Lythrine, Purity: ≥95%, CAS: 5286-10-2, 25 mg
SDP
Catalog No. 502934982
Encompass_Preferred
An alkaloid; inhibits prostaglandin synthetase (IC50 = 469 μM)
Catalog No. 50-293-4982 Supplier Cayman Chemical Supplier No. 2580425 MG
An alkaloid; inhibits prostaglandin synthetase (IC50 = 469 μM)