Learn More
Cayman Chemical Lythrine, Purity: ≥95%, CAS: 5286-10-2, 5 mg
SDP
Catalog No. 502936965
Encompass_Preferred
An alkaloid; inhibits prostaglandin synthetase (IC50 = 469 μM)
Catalog No. 50-293-6965 Supplier Cayman Chemical Supplier No. 258045 MG
An alkaloid; inhibits prostaglandin synthetase (IC50 = 469 μM)