Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hemacolor™ Rapid Staining of Blood Smear

Catalog No. M1119552500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1119552500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1119552500 Supplier MilliporeSigma™ Supplier No. 1.11955.2500
Only null left
Add to Cart
Add to Cart

Used in clinical cytology

Applications
    • For staining urine, sputum, FNAB, sperm, effusions lavages
    • For staining blood bone marrow smears
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Packaging Glass Bottle
Quantity 2.5 L
Storage Requirements 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.