Learn More
Andwin Scientific HEMACYTOMETER COVER GLASS
SDP
Catalog No. NC9995852
Encompass_Preferred
The Hemacytometer Cover Glasses are made from highly polished, hard-ground glass. This product is transparent and has rectangular edges with clipped corners. It has a dimension of 20 x 26 mm (7/8 x 1") and a thickness of 0.4 mm.
Catalog No. NC9995852 Supplier Andwin Scientific Supplier No. 16007000
The Hemacytometer Cover Glasses are made from highly polished, hard-ground glass. This product is transparent and has rectangular edges with clipped corners. It has a dimension of 20 x 26 mm (7/8 x 1") and a thickness of 0.4 mm.