Learn More
ACROBiosystems Monoclonal Anti-Influenza A [A/guinea fowl/Hong Kong/WF10/99(H9N2)] HA1 Antibody, Human IgG1 (8A1) (MALS verified)
Catalog No. 502719515
Encompass_Preferred
Immobilized Influenza A [A/guinea fowl/Hong Kong/WF10/99(H9N2)] HA1 Protein, His Tag (Cat. No. HA1-V52H5) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Influenza A [A/guinea fowl/Hong Kong/WF10/99(H9N2)] HA1 Antibody, Human IgG1 (8A1) (Cat. No. HA1-M694) with a linear range of 0.006-0.8 ng/mL (QC tested).
Catalog No. 50-271-9515 Supplier ACROBiosystems Supplier No. HA1M6941MG
Immobilized Influenza A [A/guinea fowl/Hong Kong/WF10/99(H9N2)] HA1 Protein, His Tag (Cat. No. HA1-V52H5) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Influenza A [A/guinea fowl/Hong Kong/WF10/99(H9N2)] HA1 Antibody, Human IgG1 (8A1) (Cat. No. HA1-M694) with a linear range of 0.006-0.8 ng/mL (QC tested).