Learn More
ACROBiosystems ACROBiosystems Influenza A [A/Shanghai/2/2013(H7N9)] Hemagglutinin (HA) Protein, Fc Tag
Supplier: ACROBiosystems HA9V5253100UG
Recombinant Protein;100UG;Influenza A [A/Shanghai/2/2013(H7N9)] HA, Fc Tag (HA9-V5253) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Gly 338, Leu 341 - Asp 523 (Accession # R4NN21-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 82.5 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.