Learn More
ACROBiosystems Hemagglutinin HA 100ug
Catalog No. 502534895
Encompass_Preferred
Influenza A (Vietnam/1194/2004(H5N1)) Hemagglutinin (HA) Protein His Tag (HA1-V52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 12 - Ser 529 (Accession # EPI_ISL_30632 GISAID).
Catalog No. 50-253-4895 Supplier ACROBiosystems Supplier No. HA1V52H9100UG
Influenza A (Vietnam/1194/2004(H5N1)) Hemagglutinin (HA) Protein His Tag (HA1-V52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 12 - Ser 529 (Accession # EPI_ISL_30632 GISAID).