Learn More
ACROBiosystems Hemagglutinin HA 100ug
Supplier:  ACROBiosystems HA1V52H8100UG
Encompass_Preferred
Influenza A [Victoria/4897/2022] Hemagglutinin (HA) Protein His Tag (HA1-V52H8) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - Ile 530 (Accession # EPI_ISL_17102775 GISAID).
Catalog No. 50-253-4897