Learn More
ACROBiosystems Influenza A [A/Bangkok/1/1979 (H3N2)] HA, His Tag (HA2-V52H3) is expressed from human 293 cells (HEK293). It contains AA Asn 18 - Trp 530 (Accession # AFM68976.1).
Catalog No. 502253592
Encompass_Preferred
Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Hemagglutinin also plays a major role in the determination of host range restriction and virulence. As a class I viral fusion protein, hemagglutinin is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane.
This item is not returnable. View return policy
Catalog No. 50-225-3592 Supplier ACROBiosystems Supplier No. HA2V52H31MG
Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Hemagglutinin also plays a major role in the determination of host range restriction and virulence. As a class I viral fusion protein, hemagglutinin is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane.