Learn More
ACROBiosystems Hemagglutinin HA 25ug
Catalog No. 502534886
Encompass_Preferred
Biotinylated Influenza A (A/Darwin/9/2021) Hemagglutinin (HA) Protein HisAvitag (HA2-V82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 17 - Asp 529 (Accession # EPI_ISL_2233240 GISAID).
Catalog No. 50-253-4886 Supplier ACROBiosystems Supplier No. HA2V82E625UG
Biotinylated Influenza A (A/Darwin/9/2021) Hemagglutinin (HA) Protein HisAvitag (HA2-V82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 17 - Asp 529 (Accession # EPI_ISL_2233240 GISAID).