Learn More
ACROBiosystems Influenza A (H1N1) Hemagglutinin ELISA Kit, Sandwich-ELISA, 96tests
Catalog No. 502719011
Encompass_Preferred
The Influenza A (H1N1) Viruses Hemagglutinin (HA) Specific ELISA Kit is a sandwich-ELISA based assay developed for the precise quantitative detection of Hemagglutinin (HA) from Influenza A (H1N1) viruses, including strains A/Wisconsin/588/2019, A/Victoria/4897/2022, and A/Wisconsin/67/2022, in vaccine samples. This kit is designed to provide a reliable solution for the manufacturing and quality control of HA vaccine development.
  • Quantitative detection of HA protein
  • Validated sandwich-ELISA format
  • Standard curve range of 1.563 ng/mL to 50 ng/mL
  • Validated for manufacturing and quality control applications
  • Includes pre-coated microplate, standard, HRP-conjugated antibody, and buffers
  • Storage at 2°C to 8°C
Catalog No. 50-271-9011 Supplier ACROBiosystems Supplier No. RASA18996TESTS
The Influenza A (H1N1) Viruses Hemagglutinin (HA) Specific ELISA Kit is a sandwich-ELISA based assay developed for the precise quantitative detection of Hemagglutinin (HA) from Influenza A (H1N1) viruses, including strains A/Wisconsin/588/2019, A/Victoria/4897/2022, and A/Wisconsin/67/2022, in vaccine samples. This kit is designed to provide a reliable solution for the manufacturing and quality control of HA vaccine development.
  • Quantitative detection of HA protein
  • Validated sandwich-ELISA format
  • Standard curve range of 1.563 ng/mL to 50 ng/mL
  • Validated for manufacturing and quality control applications
  • Includes pre-coated microplate, standard, HRP-conjugated antibody, and buffers
  • Storage at 2°C to 8°C