Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hemagglutinin (HA) protein, H1N1 | >95.0% | 5 UG
SDP

Catalog No. 5000290166
Encompass_Preferred

Recombinant hemagglutinin (HA0) protein from Influenza A H1N1 (A/California/04/2009), produced in Sf9 insect cells and supplied lyophilized. Intended for serological assays, antibody characterization, and vaccine research where a tag-free, native-like HA antigen is required.

  • Expressed in Sf9 insect cells, preserving eukaryotic glycosylation.
  • Tag free to reduce interference in antibody binding studies.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation for convenient storage and transport.
  • Available in small quantity options suitable for assay development.
  • Recommended reconstitution at or above 100 μg/mL; stable short-term at 4°C and long-term when frozen.

Catalog No. 50-002-90166 Supplier Medchemexpress LLC Supplier No. HYP764475UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant hemagglutinin (HA0) protein from Influenza A H1N1 (A/California/04/2009), produced in Sf9 insect cells and supplied lyophilized. Intended for serological assays, antibody characterization, and vaccine research where a tag-free, native-like HA antigen is required.

  • Expressed in Sf9 insect cells, preserving eukaryotic glycosylation.
  • Tag free to reduce interference in antibody binding studies.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation for convenient storage and transport.
  • Available in small quantity options suitable for assay development.
  • Recommended reconstitution at or above 100 μg/mL; stable short-term at 4°C and long-term when frozen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.