Learn More
Medchemexpress LLC Hemagglutinin (HA) protein, H9N2 | 97.3% | 10 UG
SDP
Catalog No. 5000293690
Encompass_Preferred
Recombinant hemagglutinin (HA) protein from influenza A H9N2, expressed in Sf9 insect cells and provided with a C-terminal His tag. Supplied as a purified antigen for serology, immunogenicity, assay development, and antibody generation.
  • Purified recombinant antigen with reported purity of 97.3%.
  • Approximate molecular weight 58.3 kDa.
  • Expressed in Sf9 insect cells with a C-terminal His tag for detection or purification.
  • Suitable for serology and antigenicity studies, and for use as an immunogen.
  • Supplied in a 10 μg vial format for small-scale assays.
Catalog No. 50-002-93690 Supplier Medchemexpress LLC Supplier No. HYP7501210UG
Recombinant hemagglutinin (HA) protein from influenza A H9N2, expressed in Sf9 insect cells and provided with a C-terminal His tag. Supplied as a purified antigen for serology, immunogenicity, assay development, and antibody generation.
  • Purified recombinant antigen with reported purity of 97.3%.
  • Approximate molecular weight 58.3 kDa.
  • Expressed in Sf9 insect cells with a C-terminal His tag for detection or purification.
  • Suitable for serology and antigenicity studies, and for use as an immunogen.
  • Supplied in a 10 μg vial format for small-scale assays.