Learn More
Medchemexpress LLC Hemagglutinin (HA) protein, Influenza A H4N2 | 96.0% | 5 UG
SDP
Catalog No. 5000291220
Encompass_Preferred
Recombinant hemagglutinin (HA) protein from Influenza A H4N2, expressed in Sf9 insect cells and purified with a C-terminal His tag. The protein is approximately 58.8 kDa with endotoxin <1 EU/μg and is supplied as a 5 μg purified antigen for use in immunoassays, binding studies, and antibody generation.
  • Recombinant influenza A H4N2 hemagglutinin protein.
  • Expressed in Sf9 insect cells for eukaryotic folding and glycosylation.
  • His tag enables affinity purification and detection.
  • Approximately 58.8 kDa molecular weight.
  • Low endotoxin level (<1 EU/μg) suitable for immunological assays.
  • Supplied as a small 5 μg aliquot for antigen and assay development.
  • Suitable for ELISA, serology, antibody production, and viral entry studies.
Catalog No. 50-002-91220 Supplier Medchemexpress LLC Supplier No. HYP750755UG
Add to Cart
Add to Cart
Recombinant hemagglutinin (HA) protein from Influenza A H4N2, expressed in Sf9 insect cells and purified with a C-terminal His tag. The protein is approximately 58.8 kDa with endotoxin <1 EU/μg and is supplied as a 5 μg purified antigen for use in immunoassays, binding studies, and antibody generation.
  • Recombinant influenza A H4N2 hemagglutinin protein.
  • Expressed in Sf9 insect cells for eukaryotic folding and glycosylation.
  • His tag enables affinity purification and detection.
  • Approximately 58.8 kDa molecular weight.
  • Low endotoxin level (<1 EU/μg) suitable for immunological assays.
  • Supplied as a small 5 μg aliquot for antigen and assay development.
  • Suitable for ELISA, serology, antibody production, and viral entry studies.