Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hemagglutinin (HA) protein, Influenza A H5N6 | >95.0% | 10 UG
SDP

Catalog No. 5000292759
Encompass_Preferred

Recombinant hemagglutinin (HA) protein from Influenza A H5N6 (A/yunnan/0127/2015), expressed in Sf9 insect cells and purified with a C-terminal His tag. Supplied as a 10 μg vial of purified protein for research use in immunoassays, structural studies, assay development, and antibody generation. Purity typically >95% by COA; consult batch datasheet for lot-specific details.

  • Recombinant full-length hemagglutinin suitable for in vitro assays.
  • Expressed in Sf9 insect cells for mammalian-like glycosylation.
  • C-terminal His tag enables affinity purification and detection.
  • Supplied as 10 μg of protein; confirm form and concentration on batch datasheet.
  • Purity greater than 95% as determined by analytical methods.
  • Intended for research use only; not for diagnostic or therapeutic use.

Catalog No. 50-002-92759 Supplier Medchemexpress LLC Supplier No. HYP7502710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant hemagglutinin (HA) protein from Influenza A H5N6 (A/yunnan/0127/2015), expressed in Sf9 insect cells and purified with a C-terminal His tag. Supplied as a 10 μg vial of purified protein for research use in immunoassays, structural studies, assay development, and antibody generation. Purity typically >95% by COA; consult batch datasheet for lot-specific details.

  • Recombinant full-length hemagglutinin suitable for in vitro assays.
  • Expressed in Sf9 insect cells for mammalian-like glycosylation.
  • C-terminal His tag enables affinity purification and detection.
  • Supplied as 10 μg of protein; confirm form and concentration on batch datasheet.
  • Purity greater than 95% as determined by analytical methods.
  • Intended for research use only; not for diagnostic or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.