Learn More
Medchemexpress LLC Hemagglutinin protein (H4N2, ADN78261) | 10 UG
SDP
Catalog No. 5000293676
Encompass_Preferred
Recombinant hemagglutinin H4 protein derived from Influenza A H4N2 (accession ADN78261). The protein is expressed in Sf9 insect cells, carries a C-terminal His tag, and is supplied as a filtered solution in 20 mM Tris, 500 mM NaCl, pH 8.0, 10% glycerol. Endotoxin is specified as <1 EU/μg. Intended for research use; aliquot and avoid repeated freeze-thaw cycles.
Catalog No. 50-002-93676 Supplier Medchemexpress LLC Supplier No. HYP7507510UG
Add to Cart
Add to Cart
Recombinant hemagglutinin H4 protein derived from Influenza A H4N2 (accession ADN78261). The protein is expressed in Sf9 insect cells, carries a C-terminal His tag, and is supplied as a filtered solution in 20 mM Tris, 500 mM NaCl, pH 8.0, 10% glycerol. Endotoxin is specified as <1 EU/μg. Intended for research use; aliquot and avoid repeated freeze-thaw cycles.