Learn More
ACROBiosystems Recombinant Protein;Recombinant Influenza A [A/Shanghai/2/2013(H7N9)] Hemagglutinin (HA), Fc Tag;HEK293;1MG;HA9,HA,Hemagglutinin
Catalog No. 501658218
Encompass_Preferred
Recombinant Protein;Recombinant Influenza A [A/Shanghai/2/2013(H7N9)] Hemagglutinin (HA), Fc Tag;1MG;Influenza , Fc Tag (HA9-V5253) is expressed from human 293 cells (HEK293).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.5 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8218 Supplier ACROBiosystems Supplier No. HA9V52531MG
Recombinant Protein;Recombinant Influenza A [A/Shanghai/2/2013(H7N9)] Hemagglutinin (HA), Fc Tag;1MG;Influenza , Fc Tag (HA9-V5253) is expressed from human 293 cells (HEK293).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.5 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.