Learn More
ACROBiosystems Recombinant Protein;Recombinant Influenza A [A/Hong Kong/483/97 (H5N1)] Hemagglutinin (HA);HEK293;1MG;HA1,HA,Hemagglutinin
Catalog No. 501658605
Encompass_Preferred
Recombinant Protein;Recombinant Influenza A [A/Hong Kong/483/97 (H5N1)] Hemagglutinin (HA);1MG;Influenza , His Tag (HA1-V5229) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Gln 531 (Accession # AAC32099.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.3 kDa. The protein migrates as 66-100 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8605 Supplier ACROBiosystems Supplier No. HA1V52291MG
Recombinant Protein;Recombinant Influenza A [A/Hong Kong/483/97 (H5N1)] Hemagglutinin (HA);1MG;Influenza , His Tag (HA1-V5229) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Gln 531 (Accession # AAC32099.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.3 kDa. The protein migrates as 66-100 kDa under reducing (R) condition (SDS-PAGE).