Learn More
Dynamic Diagnostics Hemaprep - Automated Blood Smearing Instrument
Catalog No. 22899509
Change view
Click to view available options
Product Type:
Blood Smear Slide Maker
Catalog No. Product Type
22-899-509 Blood Smear Slide Maker
1 options
Catalog No. 22-899-509 Supplier Dynamic Diagnostics Supplier No. 4000104
Add to Cart
Add to Cart
Hemaprep is the leading choice of hospital labs for automated bloodsmearing. Easy-to-use, small, self contained unit prepares smears for routine and special hematology. This unit is small enough to fit on a drawing tray and needs no additional power source.
  • Versatile: Prepares smears for routine and special hematology
  • Portable: Completely self-contained and can be carried on a blood drawing tray
  • Easy-to-use: Enables personnel with little experience to produce high quality smears
  • 4 Simple steps prepare 2 slides at the same time
  • No power source needed, trouble-free operation at a low cost (no power source needed)
  • Adjustable-Variable spreader speed allows operator to control smear length
  • 1 pair of Replacement Spreader Blades included
  • 1-year manufacturer warranty (requires registration)
  • Used in hospital laboratories for over 20 years
Specifications
Product Type Blood Smear Slide Maker
Includes 1 pair of Replacement Spreader Blades, 1 year warranty
For Use With (Application) Hematology