Learn More
Volu Sol Wright Stain, Modified, Hema-Tek™ Stain Kit, Volu-Sol™
SDP
Supplier:  Volu Sol VHA300
Encompass_Preferred
Stain pack designed for use on the Hema-Tek 1, 1000, and 2000. Compatible for use on Advia S60 Autoslide Staining System. Three (3) Reagent Pack including buffer, stain, and rinse.
Catalog No. 50-169-4709