Learn More
EUROTROL Hemotrol
Catalog No. NC1179378
Encompass
Hemotrol is a hemostatic agent used to achieve hemostasis in surgical procedures. It is composed of a plant-based chitosan derivative that activates platelets and the coagulation cascade upon contact with blood. The product comes in multiple formats including pads, powders, and liquids. Hemotrol can be used in open and laparoscopic procedures across multiple specialties including cardiovascular, orthopedic, general, plastic, and trauma surgery. It is biocompatible, resorbable, and helps reduce bleeding complications and procedure time. The website indicates Hemotrol has been used successfully in over 5 million procedures worldwide.
This item is not returnable. View return policy
Catalog No. NC1179378 Supplier EUROTROL Supplier No. 171.001.002
Hemotrol is a hemostatic agent used to achieve hemostasis in surgical procedures. It is composed of a plant-based chitosan derivative that activates platelets and the coagulation cascade upon contact with blood. The product comes in multiple formats including pads, powders, and liquids. Hemotrol can be used in open and laparoscopic procedures across multiple specialties including cardiovascular, orthopedic, general, plastic, and trauma surgery. It is biocompatible, resorbable, and helps reduce bleeding complications and procedure time. The website indicates Hemotrol has been used successfully in over 5 million procedures worldwide.