Learn More
EUROTROL Hemotrol
Catalog No. NC1179379
Encompass
Hemotrol is a hemostatic agent used to achieve hemostasis in surgery. It is composed of a plant-based chitosan derived from shrimp shells. The product comes in multiple formats including pads, packing strips, and powders. Hemotrol works by aggregating red blood cells and platelets to quickly form a strong clot. It can be used in various surgeries including cardiac, orthopedic, OB/GYN, general, reconstructive, and trauma procedures. Hemotrol is biocompatible, biodegradable, and bacteriostatic. It helps reduce blood loss and risk of infection. The product provides rapid hemostasis in 2-3 minutes.
This item is not returnable. View return policy
Catalog No. NC1179379 Supplier EUROTROL Supplier No. 171.002.002
Hemotrol is a hemostatic agent used to achieve hemostasis in surgery. It is composed of a plant-based chitosan derived from shrimp shells. The product comes in multiple formats including pads, packing strips, and powders. Hemotrol works by aggregating red blood cells and platelets to quickly form a strong clot. It can be used in various surgeries including cardiac, orthopedic, OB/GYN, general, reconstructive, and trauma procedures. Hemotrol is biocompatible, biodegradable, and bacteriostatic. It helps reduce blood loss and risk of infection. The product provides rapid hemostasis in 2-3 minutes.