Learn More
Microscope World Fein Optic RB30 Hematology Microscope
SDP
Catalog No. NC2965677
Encompass
The Fein Optic RB30 hematology microscopes are designed for blood analysis.Reversed quintuple objective turret holds the included DIN Infinity Corrected Optical System (ICOS) Objectives.Plan Achromat 10x Objective, NA 0.25, WD 12.1mmPlan Achromat 40x Objective, NA 0.65, WD 0.36mmPlan Achromat 50x Objective, NA 0.95, WD 0.19mm, Oil ImmersionPlan Achromat 100x Objective, NA 1.25, WD 0.18mm, Oil ImmersionTrinocular head. Interpupillary adjustment from 50-75mm.PL-WF 10x Widefield, plan eyepieces, FN20. Dual diopter with +/-5 adjustment. Magnification- 100x, 400x, 500x and 1000x.150x140mm mechanical stage with 76mm x 50mm movement range. Precision 0.1mm.3W LED transmitted Koehler light system. NA 1.25 Koehler illuminator condenser with slot for darkfield slider(sold separate).Coaxial coarse and fine focusing. Coarse focusing is 30mm with tightness adjustment and rack stop. Fine adjustment of 0.002mm.7.8inW x 11.6in D x 15.6inH, 22 lbs.Warranty- 5 Years.
Catalog No. NC2965677 Supplier Microscope World Supplier No. RB31HEMA
The Fein Optic RB30 hematology microscopes are designed for blood analysis.Reversed quintuple objective turret holds the included DIN Infinity Corrected Optical System (ICOS) Objectives.Plan Achromat 10x Objective, NA 0.25, WD 12.1mmPlan Achromat 40x Objective, NA 0.65, WD 0.36mmPlan Achromat 50x Objective, NA 0.95, WD 0.19mm, Oil ImmersionPlan Achromat 100x Objective, NA 1.25, WD 0.18mm, Oil ImmersionTrinocular head. Interpupillary adjustment from 50-75mm.PL-WF 10x Widefield, plan eyepieces, FN20. Dual diopter with +/-5 adjustment. Magnification- 100x, 400x, 500x and 1000x.150x140mm mechanical stage with 76mm x 50mm movement range. Precision 0.1mm.3W LED transmitted Koehler light system. NA 1.25 Koehler illuminator condenser with slot for darkfield slider(sold separate).Coaxial coarse and fine focusing. Coarse focusing is 30mm with tightness adjustment and rack stop. Fine adjustment of 0.002mm.7.8inW x 11.6in D x 15.6inH, 22 lbs.Warranty- 5 Years.