Learn More
Medmark Technologies LLC HEMATOLOGY MIXER 115V 60/HZ
Catalog No. NC0576459
Encompass_Preferred
Hematology Mixer 115 Volt. MedMark lab mixers. The original Rock & Roll Mixer rotates at 24 rpm. The air bubble uniformly wipes the tube wall clean forming complete homogenous suspension, holds 10 sample 16 x 100mm tubes or 20 small control vials.
Catalog No. NC0576459 Supplier Medmark Technologies LLC Supplier No. 140601
Hematology Mixer 115 Volt. MedMark lab mixers. The original Rock & Roll Mixer rotates at 24 rpm. The air bubble uniformly wipes the tube wall clean forming complete homogenous suspension, holds 10 sample 16 x 100mm tubes or 20 small control vials.