Learn More
Volu Sol Hematology Rinse, 32 oz / 1 L, Volu-Sol™
SDP
Catalog No. NC9610455 Shop All Volu Sol Products
Encompass_Preferred
Rinse, for use with Wright Stain, Wright-Giemsa Stain, and Hematology Buffer. Used as a final rinse solution for blood smears. The controlled pH of 7.0 and the buffering capacity allows for faster drying, superior staining results, less precipitates
Catalog No. NC9610455 Supplier Volu Sol Supplier No. VHR032
Rinse, for use with Wright Stain, Wright-Giemsa Stain, and Hematology Buffer. Used as a final rinse solution for blood smears. The controlled pH of 7.0 and the buffering capacity allows for faster drying, superior staining results, less precipitates