Learn More
Volu Sol Hematology Rinse, 5 gal / 20 L, Volu-Sol™
SDP
Supplier:  Volu Sol VHR640
Encompass_Preferred
Rinse, for use with Wright Stain, Wright-Giemsa Stain, and Hematology Buffer. Used as a final rinse solution for blood smears. The controlled pH of 7.0 and the buffering capacity allows for faster drying, superior staining results, less precipitates
Catalog No. NC9403215