Learn More
Enzo Life Sciences SCREEN-WELL Hematopoietic Toxicity library (100µl/well)
SDP
Catalog No. 502010223
Encompass_Preferred
The SCREEN-WELL Hematopoietic Toxicity Library is a focused collection of 115 compounds with defined and diverse hematopoietic toxicity, including myelosuppression, neutropenia, leukopenia, anemia, and many more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. The compounds are dissolved in DMSO at 10mM and aliquoted into deep-well plates. The library is an essential tool for predictive toxicology screening and assay development. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0223 Supplier Enzo Life Sciences Supplier No. BML28520100
The SCREEN-WELL Hematopoietic Toxicity Library is a focused collection of 115 compounds with defined and diverse hematopoietic toxicity, including myelosuppression, neutropenia, leukopenia, anemia, and many more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. The compounds are dissolved in DMSO at 10mM and aliquoted into deep-well plates. The library is an essential tool for predictive toxicology screening and assay development. Long Term Storage: -80°C. Applications: HTS .