Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hematoporphyrin monomethyl ether | 148471-91-4 | MFCD32215011 | 95.0% | 1225.43 | C70H80N8O12 | 5 MG
SDP

Catalog No. 5000223129
Encompass_Preferred

Hematoporphyrin monomethyl ether is a porphyrin-based photosensitizer used in photodynamic therapy and related biochemical research. It is supplied as a purple to purplish-red solid with high purity and characterized solubility in DMSO; handle and store protected from light.

  • Solid, purple to purplish red appearance
  • Purity approximately 95.0%
  • Soluble in DMSO (≈62.5 mg/mL); in vivo formulations documented
  • Storage: solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month); protect from light
  • Used as a photosensitizer for photodynamic therapy research and tissue visualization

Catalog No. 50-002-23129 Supplier Medchemexpress LLC Supplier No. HY1349905MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hematoporphyrin monomethyl ether is a porphyrin-based photosensitizer used in photodynamic therapy and related biochemical research. It is supplied as a purple to purplish-red solid with high purity and characterized solubility in DMSO; handle and store protected from light.

  • Solid, purple to purplish red appearance
  • Purity approximately 95.0%
  • Soluble in DMSO (≈62.5 mg/mL); in vivo formulations documented
  • Storage: solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month); protect from light
  • Used as a photosensitizer for photodynamic therapy research and tissue visualization

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.