Learn More
Medchemexpress LLC Hematoporphyrin monomethyl ether | 148471-91-4 | MFCD32215011 | 95.0% | C70H80N8O12 | 10MG
SDP
Catalog No. 5000260939
Encompass_Preferred
Hematoporphyrin monomethyl ether is a porphyrin-related photosensitizer used as a research reagent for photodynamic studies. It produces singlet oxygen when activated by light and is commonly applied in photodynamic therapy research, cellular photochemistry, and light-activated imaging experiments.
  • Porphyrin-derived photosensitizer with high singlet oxygen yield.
  • Suitable for photodynamic therapy research and in vitro photochemistry.
  • Solid with a purple to purplish red appearance for easy visual identification.
  • Available in small research pack sizes suitable for laboratory experiments.
  • Store protected from light and refrigerate to maintain stability.
Catalog No. 50-002-60939 Supplier Medchemexpress LLC Supplier No. HY13499010MG
Hematoporphyrin monomethyl ether is a porphyrin-related photosensitizer used as a research reagent for photodynamic studies. It produces singlet oxygen when activated by light and is commonly applied in photodynamic therapy research, cellular photochemistry, and light-activated imaging experiments.
  • Porphyrin-derived photosensitizer with high singlet oxygen yield.
  • Suitable for photodynamic therapy research and in vitro photochemistry.
  • Solid with a purple to purplish red appearance for easy visual identification.
  • Available in small research pack sizes suitable for laboratory experiments.
  • Store protected from light and refrigerate to maintain stability.