Learn More
Cayman Chemical Hematoporphyrin (hydrochloride), Purity: ≥90% (mixture of isomers), CAS: 17696-69-4, 250 mg
SDP
Catalog No. 502937382
Encompass_Preferred
A photosensitizer; increases oxygen consumption and decreases the RCR in irradiated isolated rat liver mitochondria at 3 μM; It induces DNA breaks in cell-free assays at 6 μM; decreases the survival of irradiated HeLa cells at 12 μM; decreases growth of subcutaneous Yoshida AH-130 hepatoma tumors in rats when administered with radiation at 5 mg/kg
Catalog No. 50-293-7382 Supplier Cayman Chemical Supplier No. 28287250 MG
A photosensitizer; increases oxygen consumption and decreases the RCR in irradiated isolated rat liver mitochondria at 3 μM; It induces DNA breaks in cell-free assays at 6 μM; decreases the survival of irradiated HeLa cells at 12 μM; decreases growth of subcutaneous Yoshida AH-130 hepatoma tumors in rats when administered with radiation at 5 mg/kg