Learn More
STEMCELL Technologies HemaTox™ Erythroid Kit, Size: 1 Kit
SDP
Catalog No. NC1593313
Encompass
Serum-free medium and supplement for the measurement of drug toxicity on erythroid cells
This item is not returnable. View return policy
Catalog No. NC1593313 Supplier STEMCELL Technologies Supplier No. 09701
Serum-free medium and supplement for the measurement of drug toxicity on erythroid cells