Learn More
Bioworld Hematoxylin, 25 g
Catalog No. 50199219
Encompass_Preferred
A general histological stain. High Purity Grade EM (550nm, 0.1M NaOH) >14,700
Catalog No. 50-199-219 Supplier Bioworld Supplier No. 217700231
A general histological stain. High Purity Grade EM (550nm, 0.1M NaOH) >14,700