Learn More
Bioworld Hematoxylin, 100 g
Catalog No. 50199221
Encompass_Preferred
A general histological stain. High Purity Grade EM (550nm, 0.1M NaOH) >14,700
Catalog No. 50-199-221 Supplier Bioworld Supplier No. 217700233
A general histological stain. High Purity Grade EM (550nm, 0.1M NaOH) >14,700