Learn More
Diagnostic Biosystems HEMATOXYLIN Montage™ Hematoxylin
Catalog No. 502407596
Encompass_Preferred
Hematoxylin is a histological staining reagent suitable for visualization of nuclei in tissue sections. Montage™ Hematoxylin, is designed for optimal performance when used together with the Montage 360™ System.\n\nDBS’s Hematoxylin is intended for use in the histologic demonstration of nuclear staining. This staining technique is used to make the critical distinction between a normal nucleus and an abnormal one.
Catalog No. 50-240-7596 Supplier Diagnostic Biosystems Supplier No. K056100AN
Hematoxylin is a histological staining reagent suitable for visualization of nuclei in tissue sections. Montage™ Hematoxylin, is designed for optimal performance when used together with the Montage 360™ System.\n\nDBS’s Hematoxylin is intended for use in the histologic demonstration of nuclear staining. This staining technique is used to make the critical distinction between a normal nucleus and an abnormal one.