Learn More
Alkali Scientific Hematoxylin, 25 G
SDP
Catalog No. 5000659692
Encompass_Preferred
Hematoxylin, 25g, is a commonly used histological stain for visualizing cell structures, especially cell nuclei, during tissue analysis. The staining process is essential in histology, allowing pathologists and researchers to distinguish between different tissue components. Hematoxylin provides clear and consistent staining of cell nuclei, turning them blue or purple, which makes it a key reagent in tissue examination. This 25g quantity is perfect for smaller labs or research projects, offering an economical solution for routine staining procedures. Hematoxylin is especially useful in combination with eosin for Hematoxylin and Eosin (H&E) staining, the most widely used tissue stain in medical research.
Catalog No. 50-006-59692 Supplier Alkali Scientific Supplier No. HEM002CP
Hematoxylin, 25g, is a commonly used histological stain for visualizing cell structures, especially cell nuclei, during tissue analysis. The staining process is essential in histology, allowing pathologists and researchers to distinguish between different tissue components. Hematoxylin provides clear and consistent staining of cell nuclei, turning them blue or purple, which makes it a key reagent in tissue examination. This 25g quantity is perfect for smaller labs or research projects, offering an economical solution for routine staining procedures. Hematoxylin is especially useful in combination with eosin for Hematoxylin and Eosin (H&E) staining, the most widely used tissue stain in medical research.