Learn More
Leica Biosystems HEMATOXYLIN 25ML
Catalog No. NC0789326
Encompass_Preferred
Novocastra Hematoxylin, RE7107, is intended for use in immunohistochemical (IHC) staining procedures. Hematoxylin stains cell nuclei and has many uses in histology, the most common of which is the Hematoxylin and Eosin stain. In IHC procedures, hematoxylin can be used as a counterstain to aid the visualization and localization of the colored end product. 25 mL of the reagent is supplied.
Catalog No. NC0789326 Supplier Leica Biosystems Supplier No. RE7107CE
Novocastra Hematoxylin, RE7107, is intended for use in immunohistochemical (IHC) staining procedures. Hematoxylin stains cell nuclei and has many uses in histology, the most common of which is the Hematoxylin and Eosin stain. In IHC procedures, hematoxylin can be used as a counterstain to aid the visualization and localization of the colored end product. 25 mL of the reagent is supplied.