Learn More
Novus Biologicals™ Hematoxylin

Description
Biological Activity: Dye used to stain cell nuclei
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Description | CAS: 517-28-2 |
| Formulation | C16H14O6 |
| For Use With (Application) | Used in stain nuclei |
| Quantity | 25 g |
| Product Type | Dye |
| Molecular Weight (g/mol) | 302.28g/mol |
| Solubility | Soluble to 3mM in ethanol |
| Content And Storage | Room Temperature |
| Target | Hematoxylin |
| Color | Beige |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.