Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Hematoxylin, 85%, pure, certified

Catalog No. AC411160250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411160250 25 g Glass Bottle
AC411161000 100 g Glass Bottle
AC411165000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411160250 Supplier Thermo Scientific Chemicals Supplier No. 411160250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 517-28-2 | C16H14O6 | 302.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black 1, C.I. 75290
PubChem CID 45029742
IUPAC Name (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

Melting Point 100.0°C to 120.0°C
Infrared Spectrum Authentic
Loss on Drying 10% (105°C) max.
Packaging Glass Bottle
Assay Percent Range 85%
UV_Vis 88% (on dry substance) min.
Color Beige to Brown
Solubility Information Solubility in water: slightly soluble; soluble in hot water;. Other solubilities: soluble in hot alcohol, glycerol, borax, alkali hy, droxides (aq. solution purple red; becomes blue vi, olett during storage at air).
Quantity 25 g
Physical Form Powder
Formula Weight 302.27
Percent Purity 85%
Grade Pure
Chemical Name or Material Hematoxylin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.