Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Hematoxylin, Certified, C.I. DcH-48 10 GR
SDP

Catalog No. 5026465
Encompass_Preferred

(Natural Black I, Hydroxybrasilin)
CAS #517-28-2 C₁₆H₁₄O₆
Solubility: 10.0% Water; 10.0% Alcohol; 9.5% Cellosolve; 10% Glycol; 0.0% Xylene

A certified biological stain used in histology and cytology and plastic embedded tissues. See Chen, Chang, S., J Arch. of Path., 93,334 (1972).

Catalog No. 50-264-65 Supplier Electron Microscopy Sciences Supplier No. 16620
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Natural Black I, Hydroxybrasilin)
CAS #517-28-2 C₁₆H₁₄O₆
Solubility: 10.0% Water; 10.0% Alcohol; 9.5% Cellosolve; 10% Glycol; 0.0% Xylene

A certified biological stain used in histology and cytology and plastic embedded tissues. See Chen, Chang, S., J Arch. of Path., 93,334 (1972).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.