Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematoxylin (Delafields), Vion Biosciences
SCI_ED

Catalog No. S02498 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02498 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm.
Catalog No. S02498 Supplier Aldon Corporation Supplier No. VNHH0012500ML
Add to Cart
Add to Cart
WARNING: This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm.

Use hematoxylin in tandem with Eosin Y or to visualize DNA and RNA in histological samples.

  • Use hematoxylin in tandem with Eosin Y or to visualize DNA and RNA in histological samples.

Specifications

Boiling Point Data not available.
CAS 7732-18-5
Certifications/Compliance What standards of certification or compliance does the product meet?
Color Dark Red-Purple
Compatible Cells Organ and Tissue Cells
Concentration 0.70%
Description Hematoxylin Solution (Delafield's)
Detection Method H&E Staining
Form Liquid
For Use With (Application) Histology
Label or Dye Dye
Melting Point Data not available.
Molecular Formula Mixture
Packaging HDPE Bottle
pH Data not available.
Product Line Vion Histology
Purity 0.70%
Quantity 500 mL
Reagent Type Stain
Sample Type Tissue
Solubility Complete in water.
Solution Type Alcoholic
Storage Requirements Store in a cool, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Sub Cellular Localization Nuclei - DNA, RNA, chromatin
Synonym Delafield's Hematoxylin Solution
Target Species Human
Product Type Hematoxylin (Delafields)
Assay 0.70%
Show More Show Less
ShelfLife 3 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.