Learn More
Vector Laboratories Hematoxylin and Eosin Stain Kit
SDP
Catalog No. NC1470670
Encompass
The Vector Laboratories Hematoxylin and Eosin Stain Kit is intended for use in histology and cytology applications. Included in this kit is a modified Eosin that provides the benefits of a traditional alcoholic formulation with significant improvements in usability. Advantages include lower evaporation rate, better color patterns, and improved surface tension to remain on tissue section. Our Hematoxylin produces crisp, intense blue nuclei providing optimal contrast to the Eosin stained cytoplasm.
Catalog No. NC1470670 Supplier Vector Laboratories Supplier No. H3502
The Vector Laboratories Hematoxylin and Eosin Stain Kit is intended for use in histology and cytology applications. Included in this kit is a modified Eosin that provides the benefits of a traditional alcoholic formulation with significant improvements in usability. Advantages include lower evaporation rate, better color patterns, and improved surface tension to remain on tissue section. Our Hematoxylin produces crisp, intense blue nuclei providing optimal contrast to the Eosin stained cytoplasm.