Learn More
Scytek Laboratories Hematoxylin and Eosin Stain Kit
SDP
Catalog No. NC0510871
Encompass_Preferred
The Hematoxylin and Eosin Stain Kit is intended for use in histology and cytology applications. Included in this kit is a newly formulated Eosin that provides the benefits of a traditional alcoholic formula with significant improvements in usability. Advantages include lower evaporation rate, better color patterns, reduced tendency to spill over container, hands, and countertops, and improved surface tension to remain on tissue section. Our Hematoxylin produces crisp, intense blue nuclei providing optimal contrast to the Eosin stained cytoplasm. Cytoplasm: Light Pink Collagen: Pink Muscle: Pink/Rose Erythrocytes: Pink/Red Nuclei: Blue
Catalog No. NC0510871 Supplier Scytek Laboratories Supplier No. HAE1
The Hematoxylin and Eosin Stain Kit is intended for use in histology and cytology applications. Included in this kit is a newly formulated Eosin that provides the benefits of a traditional alcoholic formula with significant improvements in usability. Advantages include lower evaporation rate, better color patterns, reduced tendency to spill over container, hands, and countertops, and improved surface tension to remain on tissue section. Our Hematoxylin produces crisp, intense blue nuclei providing optimal contrast to the Eosin stained cytoplasm. Cytoplasm: Light Pink Collagen: Pink Muscle: Pink/Rose Erythrocytes: Pink/Red Nuclei: Blue