Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematoxylin, Fisher BioReagents

Catalog No. BP2424100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2424100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP2424100

missing translation for 'validMainframeMsg'

Hematoxylin is used as a stain in microscopy.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black I
PubChem CID 45029742
IUPAC Name (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

Melting Point 100°C
Bio. Stain Comm. Certification Pass Test
Merck Index 15, 4672
Formula Weight 302.28
Packaging Glass Bottle
Color Index No. 75290
Color Beige
Quantity 100 g
Physical Form Powder
Chemical Name or Material Hematoxylin
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.