Learn More
Scytek Laboratories Hematoxylin, for Automation, 1 L
SDP
Catalog No. NC1822443
Encompass_Preferred
Hematoxylin, For Automation is ideal for use with autostainers. Hematoxylin, For Automation produces a crisp blue nuclei . Hematoxylin is a nuclear stain that is used as a counterstain for immunohistochemistry (IHC). Together with eosin, it forms the Hematoxylin and Eosin stain (H&E stain), one of the most commonly used stains in histology.
This item is not returnable. View return policy
Catalog No. NC1822443 Supplier Scytek Laboratories Supplier No. HAQ999
Hematoxylin, For Automation is ideal for use with autostainers. Hematoxylin, For Automation produces a crisp blue nuclei . Hematoxylin is a nuclear stain that is used as a counterstain for immunohistochemistry (IHC). Together with eosin, it forms the Hematoxylin and Eosin stain (H&E stain), one of the most commonly used stains in histology.