Learn More
Selleck Chemical LLC Hematoxylin 25mg 517-28-2 Hydroxybrazilin
SDP
Catalog No. 505449
Encompass_Preferred
Hematoxylin (Hydroxybrazilin) is a compound that forms strongly colored complexes with certain metal ions, notably Fe(III) and Al(III) salts and a kind of stain in histology. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-544-9 Supplier Selleck Chemical LLC Supplier No. S238425MG
Hematoxylin (Hydroxybrazilin) is a compound that forms strongly colored complexes with certain metal ions, notably Fe(III) and Al(III) salts and a kind of stain in histology. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.