Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Hematoxylin, iCON™ Staining System
SDP

Catalog No. 21377132
Change view
Click to view available options
Quantity:
16 oz.
1 gal.
4 x 16 oz.
4 x 1 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-377-132 4 x 16 oz.
21-377-129 16 oz.
21-377-130 1 gal.
21-377-131 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 21-377-132 Supplier Azer Scientific Supplier No. ES365114X16
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemically engineered to produce quality results on a consistent basis with crisp, delineated detail.

  • Produce quality results on a consistent basis with crisp, delineated detail
  • Can be used on all automatic stainers and used in manual staining
  • Designed to be used as a progressive or regressive nuclear stain
  • Contains no mercury
  • Shelf life of eighteen month at room temperature
  • This hematoxylin can be used on all automatic stainers and used in manual staining.
  • Nuclear staining intensity and density can be increased or decreased progressively by increasing or decreasing staining time by 1/2 minute intervals.

Specifications

Quantity 4 x 16 oz.
Product Type Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.