Learn More
Scytek Laboratories HEMATOXYLIN, MAYER'S, 500 ML
SDP
Catalog No. NC2026315
Encompass_Preferred
Mayer's Hematoxylin (Lillie's Modification) is a progressive nuclear hematoxylin stain with several histological applications. Nuclei should staining is strong, clean, crisp, and no differentiation is needed. Bluing reagent (Catalog: BRT) may be used following hematoxylin to ""blue"" or alter the shade of hematoxylin from a purple to blue. Nuclei: Purple. Nuclei after Bluing: Blue. Blue nuclei after Eosin: Blue to Violet.Hematoxylin is a nuclear stain that is used as a counterstain for immunohistochemistry (IHC). Together with eosin, it forms the Hematoxylin and Eosin stain (H&E stain), one of the most commonly used stains in histology.Summary Protocol:Apply Hematoxylin to completely cover specimen and incubate for 2-10 minutes with hematoxylin according to desired intensity.
Catalog No. NC2026315 Supplier Scytek Laboratories Supplier No. HMM500
Mayer's Hematoxylin (Lillie's Modification) is a progressive nuclear hematoxylin stain with several histological applications. Nuclei should staining is strong, clean, crisp, and no differentiation is needed. Bluing reagent (Catalog: BRT) may be used following hematoxylin to ""blue"" or alter the shade of hematoxylin from a purple to blue. Nuclei: Purple. Nuclei after Bluing: Blue. Blue nuclei after Eosin: Blue to Violet.Hematoxylin is a nuclear stain that is used as a counterstain for immunohistochemistry (IHC). Together with eosin, it forms the Hematoxylin and Eosin stain (H&E stain), one of the most commonly used stains in histology.Summary Protocol:Apply Hematoxylin to completely cover specimen and incubate for 2-10 minutes with hematoxylin according to desired intensity.