Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hematoxylin Monohydrate

Catalog No. M1159380100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1159380100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1159380100 Supplier MilliporeSigma™ Supplier No. 1159380100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For specific staining of DNA in the nuclei

Applications
    • Used in cytology histology
    • Nuclei are stained blue, dark violet to black
    • Hematoxylins are mixed with trivalent positively charged metal salts
    • Used for selective staining of nuclei
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
Quantity 100 g
Storage Requirements 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.