Learn More
Anatech Ltd HEMATOXYLIN NORMAL/1GAL
Catalog No. NC9937161
Encompass_Preferred
HEMATOXYLIN-NORMAL,1/gal,components:Aluminum sulfate,Ethylene glycol,Glacial acetic acid,Hematoxylin,Sodium iodate,BP:273degreeF,Evaporation rate:<1,Vapor density:> 1,Specific gravity:1.040,FP:> 200degreeF,completely soluble water,Dark purple
Catalog No. NC9937161 Supplier Anatech Ltd Supplier No. 812
HEMATOXYLIN-NORMAL,1/gal,components:Aluminum sulfate,Ethylene glycol,Glacial acetic acid,Hematoxylin,Sodium iodate,BP:273degreeF,Evaporation rate:<1,Vapor density:> 1,Specific gravity:1.040,FP:> 200degreeF,completely soluble water,Dark purple